Assay Details
| Gene Symbol: HIF1AN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009010157.2 | XP_009008405.1 | 4 - 5 | 773 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIF1AN| Gene Symbol: HIF1AN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009010157.2 | XP_009008405.1 | 4 - 5 | 773 | 66 | |
Set Membership: |
|