Assay Details
Gene Symbol: MRPL20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002750182.4 | XP_002750228.1 | 2 - 3 | 375 | 70 | |
XM_009000231.2 | XP_008998479.1 | 2 - 3 | 564 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
