Assay Details
Gene Symbol: RBAK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002744086.3 | XP_002744132.2 | 2 - 3 | 326 | 81 | |
XM_017966969.1 | XP_017822458.1 | 1 - 2 | 544 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002744086.3, XM_017966969.1 transcripts.
Set Membership: |
![]() ![]() |
