Assay Details
Gene Symbol: APITD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204928.1 | NP_001191857.1 | 3 - 4 | 611 | 96 | |
XM_009000308.2 | XP_008998556.1 | 3 - 4 | 436 | 96 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
