Assay Details
| Gene Symbol: DLEU7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002742655.3 | XP_002742701.1 | 1 | 304 | 103 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002742655.3 transcripts.
Set Membership: |
|

DLEU7
Genomic Map