Assay Details
| Gene Symbol: CCZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744061.3 | XP_002744107.1 | 12 - 13 | 1673 | 102 | |
| GenBank mRNA | EF214957.1 | - | 1 - 2 | 99 | 102 | |
More Information
Set Membership: |
|

CCZ1
Genomic Map