Assay Details
| Gene Symbol: NYAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744042.3 | XP_002744088.2 | 6 - 7 | 2464 | 113 | |
| XM_017966398.1 | XP_017821887.1 | 6 - 7 | 2468 | 113 | ||
More Information
Set Membership: |
|

NYAP1
Genomic Map