Assay Details
| Gene Symbol: ZNF771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756065.3 | XP_002756111.1 | 2 - 3 | 437 | 70 | |
| XM_009009436.2 | XP_009007684.1 | 2 - 3 | 758 | 70 | ||
| XM_009009437.2 | XP_009007685.1 | 2 - 3 | 461 | 70 | ||
More Information
Set Membership: |
|

ZNF771
Genomic Map