Assay Details
| Gene Symbol: DCTN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743035.3 | XP_002743081.1 | 1 - 2 | 151 | 80 | |
| XM_008998599.2 | XP_008996847.1 | 1 - 2 | 151 | 80 | ||
| XM_008998604.2 | XP_008996852.1 | 1 - 2 | 151 | 80 | ||
More Information
Set Membership: |
|

DCTN3
Genomic Map