Assay Details
Gene Symbol: CD55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002760749.4 | XP_002760795.2 | 5 - 6 | 952 | 58 | |
XM_008985589.2 | XP_008983837.1 | 5 - 6 | 952 | 58 | ||
XM_017966621.1 | XP_017822110.1 | 5 - 6 | 952 | 58 | ||
XM_017966622.1 | XP_017822111.1 | 5 - 6 | 952 | 58 | ||
XM_017966623.1 | XP_017822112.1 | 5 - 6 | 952 | 58 | ||
XM_017966624.1 | XP_017822113.1 | 5 - 6 | 952 | 58 | ||
XM_017966626.1 | XP_017822115.1 | 5 - 6 | 952 | 58 | ||
XM_017966627.1 | XP_017822116.1 | 5 - 6 | 952 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
