Assay Details
| Gene Symbol: ZNF45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008988148.2 | XP_008986396.1 | 4 - 5 | 532 | 62 | |
| XM_017967476.1 | XP_017822965.1 | 2 - 3 | 698 | 62 | ||
| XM_017967477.1 | XP_017822966.1 | 3 - 4 | 448 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008988148.2, XM_017967476.1, XM_017967477.1 transcripts.
Set Membership: |
|

ZNF45
Genomic Map