Assay Details
| Gene Symbol: ZNF45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008988148.2 | XP_008986396.1 | 6 - 7 | 754 | 96 | |
| XM_008988149.2 | XP_008986397.1 | 5 - 6 | 627 | 96 | ||
| XM_017967476.1 | XP_017822965.1 | 4 - 5 | 920 | 96 | ||
| XM_017967477.1 | XP_017822966.1 | 5 - 6 | 670 | 96 | ||
| XM_017967478.1 | XP_017822967.1 | 2 - 3 | 1164 | 96 | ||
More Information
Set Membership: |
|

ZNF45
Genomic Map