Assay Details
| Gene Symbol: ZNF45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008988148.2 | XP_008986396.1 | 7 | 1336 | 123 | |
| XM_008988149.2 | XP_008986397.1 | 6 | 1209 | 123 | ||
| XM_017967476.1 | XP_017822965.1 | 5 | 1502 | 123 | ||
| XM_017967477.1 | XP_017822966.1 | 6 | 1252 | 123 | ||
| XM_017967478.1 | XP_017822967.1 | 3 | 1746 | 123 | ||
More Information
Set Membership: |
|

ZNF45
Genomic Map