Assay Details
Gene Symbol: SAR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002756275.4 | XP_002756321.1 | 5 - 6 | 482 | 73 | |
XM_009009804.2 | XP_009008052.1 | 6 - 7 | 546 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
