Assay Details
| Gene Symbol: TMEM41B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009007646.2 | XP_009005894.1 | 1 - 2 | 430 | 70 | |
| XM_009007647.2 | XP_009005895.1 | 2 - 3 | 217 | 70 | ||
More Information
Set Membership: |
|

TMEM41B
Genomic Map