Assay Details
| Gene Symbol: TMX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753899.4 | XP_002753945.1 | 1 - 2 | 286 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMX1| Gene Symbol: TMX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753899.4 | XP_002753945.1 | 1 - 2 | 286 | 85 | |
Set Membership: |
|