Assay Details
| Gene Symbol: ARSF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762591.4 | XP_002762637.2 | 4 - 5 | 887 | 77 | |
| XM_008988894.2 | XP_008987142.1 | 4 - 5 | 1045 | 77 | ||
| XM_008988896.2 | XP_008987144.1 | 3 - 4 | 300 | 77 | ||
More Information
Set Membership: |
|

ARSF
Genomic Map