Assay Details
| Gene Symbol: ARSF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762591.4 | XP_002762637.2 | 9 - 10 | 1872 | 72 | |
| XM_008988894.2 | XP_008987142.1 | 9 - 10 | 2030 | 72 | ||
| XM_008988896.2 | XP_008987144.1 | 8 - 9 | 1285 | 72 | ||
| XM_008988898.2 | XP_008987146.1 | 8 - 9 | 1905 | 72 | ||
| XM_008988900.2 | XP_008987148.1 | 7 - 8 | 1754 | 72 | ||
More Information
Set Membership: |
|

ARSF
Genomic Map