Assay Details
| Gene Symbol: ZGRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008992812.2 | XP_008991060.1 | 24 - 25 | 5669 | 69 | |
| XM_008992814.2 | XP_008991062.1 | 24 - 25 | 5647 | 69 | ||
| XM_008992816.2 | XP_008991064.1 | 24 - 25 | 5642 | 69 | ||
| XM_008992817.2 | XP_008991065.1 | 24 - 25 | 5667 | 69 | ||
| XM_008992821.2 | XP_008991069.1 | 24 - 25 | 5672 | 69 | ||
| XM_008992822.2 | XP_008991070.1 | 24 - 25 | 5671 | 69 | ||
| XM_008992825.2 | XP_008991073.1 | 24 - 25 | 5070 | 69 | ||
| XM_008992826.2 | XP_008991074.1 | 24 - 25 | 5043 | 69 | ||
| XM_017970314.1 | XP_017825803.1 | 20 - 21 | 5225 | 69 | ||
More Information
Set Membership: |
|

ZGRF1
Genomic Map