Assay Details
| Gene Symbol: TTC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009006264.2 | XP_009004512.1 | 2 - 3 | 1094 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TTC9| Gene Symbol: TTC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009006264.2 | XP_009004512.1 | 2 - 3 | 1094 | 76 | |
Set Membership: |
|