Assay Details
| Gene Symbol: TEFM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748338.4 | XP_002748384.3 | 1 - 2 | 890 | 72 | |
| XM_017972543.1 | XP_017828032.1 | 1 - 2 | 898 | 72 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TEFM| Gene Symbol: TEFM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748338.4 | XP_002748384.3 | 1 - 2 | 890 | 72 | |
| XM_017972543.1 | XP_017828032.1 | 1 - 2 | 898 | 72 | ||