Assay Details
| Gene Symbol: TEFM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748338.4 | XP_002748384.3 | 2 - 3 | 1351 | 88 | |
| XM_017972543.1 | XP_017828032.1 | 2 - 3 | 1359 | 88 | ||
| XM_017972544.1 | XP_017828033.1 | 2 - 3 | 480 | 88 | ||
More Information
Set Membership: |
|

TEFM
Genomic Map