Assay Details
| Gene Symbol: TEFM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748338.4 | XP_002748384.3 | 3 - 4 | 1493 | 99 | |
| XM_017972544.1 | XP_017828033.1 | 3 - 4 | 622 | 99 | ||
| XM_017972545.1 | XP_017828034.1 | 2 - 3 | 431 | 99 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017972545.1 transcripts.
Set Membership: |
|

TEFM
Genomic Map