Assay Details
| Gene Symbol: MOSPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002763286.3 | XP_002763332.1 | 3 - 4 | 500 | 66 | |
| XM_008989939.1 | XP_008988187.1 | 3 - 4 | 500 | 66 | ||
| XM_008989940.2 | XP_008988188.1 | 3 - 4 | 500 | 66 | ||
| XM_017968663.1 | XP_017824152.1 | 3 - 4 | 500 | 66 | ||
More Information
Set Membership: |
|

MOSPD1
Genomic Map