Assay Details
| Gene Symbol: TMEM45B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754589.4 | XP_002754635.1 | 2 - 3 | 264 | 63 | |
| XM_009007103.2 | XP_009005351.1 | 2 - 3 | 257 | 63 | ||
| XM_009007104.2 | XP_009005352.1 | 2 - 3 | 280 | 63 | ||
| XM_017977642.1 | XP_017833131.1 | 2 - 3 | 263 | 63 | ||
More Information
Set Membership: |
|

TMEM45B
Genomic Map