Assay Details
| Gene Symbol: LOC100398762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009009485.1 | XP_009007733.1 | 2197 | 151 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100398762| Gene Symbol: LOC100398762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009009485.1 | XP_009007733.1 | 2197 | 151 | ||