Assay Details
| Gene Symbol: TEX35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017966047.1 | XP_017821536.1 | 6 - 7 | 680 | 80 | |
| XM_017966048.1 | XP_017821537.1 | 5 - 6 | 459 | 80 | ||
| XM_017966049.1 | XP_017821538.1 | 6 - 7 | 716 | 80 | ||
| XM_017966052.1 | XP_017821541.1 | 6 - 7 | 667 | 80 | ||
| XM_017966053.1 | XP_017821542.1 | 6 - 7 | 684 | 80 | ||
| XM_017966054.1 | XP_017821543.1 | 6 - 7 | 673 | 80 | ||
| XM_017966055.1 | XP_017821544.1 | 6 - 7 | 673 | 80 | ||
| XM_017966056.1 | XP_017821545.1 | 6 - 7 | 673 | 80 | ||
| XM_017966057.1 | XP_017821546.1 | 6 - 7 | 733 | 80 | ||
| XM_017966058.1 | XP_017821547.1 | 6 - 7 | 706 | 80 | ||
| XM_017966059.1 | XP_017821548.1 | 6 - 7 | 733 | 80 | ||
| XM_017966060.1 | XP_017821549.1 | 6 - 7 | 673 | 80 | ||
| XM_017966061.1 | XP_017821550.1 | 6 - 7 | 706 | 80 | ||
| XM_017966065.1 | XP_017821554.1 | 6 - 7 | 720 | 80 | ||
| XM_017966066.1 | XP_017821555.1 | 6 - 7 | 721 | 80 | ||
| XM_017966067.1 | XP_017821556.1 | 6 - 7 | 721 | 80 | ||
More Information
Set Membership: |
|

TEX35
Genomic Map