Assay Details
| Gene Symbol: B3GAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009007174.2 | XP_009005422.1 | 3 - 4 | 417 | 100 | |
| XM_009007176.2 | XP_009005424.1 | 3 - 4 | 381 | 100 | ||
| XM_017977667.1 | XP_017833156.1 | 3 - 4 | 417 | 100 | ||
More Information
Set Membership: |
|

B3GAT1
Genomic Map