Assay Details
| Gene Symbol: ISCA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754109.4 | XP_002754155.1 | 1 - 2 | 135 | 65 | |
| XM_009006340.1 | XP_009004588.1 | 1 - 2 | 161 | 65 | ||
| XM_009006341.2 | XP_009004589.1 | 1 - 2 | 245 | 65 | ||
| XM_017977221.1 | XP_017832710.1 | 1 - 2 | 217 | 65 | ||
More Information
Set Membership: |
|

ISCA2
Genomic Map