Assay Details
| Gene Symbol: NOSIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008988410.2 | XP_008986658.1 | 3 - 4 | 267 | 55 | |
| XM_008988411.2 | XP_008986659.1 | 3 - 4 | 200 | 55 | ||
| XM_008988412.2 | XP_008986660.1 | 3 - 4 | 263 | 55 | ||
| XM_017967974.1 | XP_017823463.1 | 3 - 4 | 300 | 55 | ||
More Information
Set Membership: |
|

NOSIP
Genomic Map