Assay Details
| Gene Symbol: FSIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017976931.1 | XP_017832420.1 | 5 - 6 | 693 | 68 | |
| XM_017976932.1 | XP_017832421.1 | 5 - 6 | 693 | 68 | ||
More Information
Set Membership: |
|

FSIP1
Genomic Map