Assay Details
| Gene Symbol: LIMD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748136.4 | XP_002748182.1 | 4 - 5 | 420 | 65 | |
| XM_008996878.2 | XP_008995126.1 | 4 - 5 | 411 | 65 | ||
More Information
Set Membership: |
|

LIMD2
Genomic Map