Assay Details
Gene Symbol: CAJA-DRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270911.1 | NP_001257840.1 | 2 - 3 | 430 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
