Assay Details
| Gene Symbol: ZNF425 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003733632.3 | XP_003733680.1 | 3 - 4 | 388 | 91 | |
| XM_009002869.2 | XP_009001117.1 | 3 - 4 | 332 | 91 | ||
More Information
Set Membership: |
|

ZNF425
Genomic Map