Assay Details
| Gene Symbol: SHISA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758310.3 | XP_002758356.2 | 4 - 5 | 578 | 99 | |
| XM_008981596.2 | XP_008979844.1 | 2 - 3 | 430 | 99 | ||
| XM_017964886.1 | XP_017820375.1 | 3 - 4 | 429 | 99 | ||
| XM_017964887.1 | XP_017820376.1 | 3 - 4 | 413 | 99 | ||
More Information
Set Membership: |
|

SHISA5
Genomic Map