Assay Details
| Gene Symbol: VWA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750195.4 | XP_002750241.2 | 1 - 2 | 1081 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

VWA1| Gene Symbol: VWA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750195.4 | XP_002750241.2 | 1 - 2 | 1081 | 101 | |
Set Membership: |
|