Assay Details
| Gene Symbol: JAML | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754476.4 | XP_002754522.1 | 9 - 10 | 1179 | 86 | |
| XM_009006908.2 | XP_009005156.1 | 8 - 9 | 1125 | 86 | ||
More Information
Set Membership: |
|

JAML
Genomic Map