Assay Details
| Gene Symbol: SLX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009009057.2 | XP_009007305.1 | 5 - 6 | 2187 | 62 | |
| XM_009009059.2 | XP_009007307.1 | 5 - 6 | 1834 | 62 | ||
| XM_009009060.2 | XP_009007308.1 | 5 - 6 | 2139 | 62 | ||
More Information
Set Membership: |
|

SLX4
Genomic Map