Assay Details
| Gene Symbol: TRABD2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008980420.1 | XP_008978668.1 | 2 | 786 | 90 | |
| XM_008980421.1 | XP_008978669.1 | 2 | 688 | 90 | ||
| XM_017964208.1 | XP_017819697.1 | 2 | 688 | 90 | ||
| XM_017964209.1 | XP_017819698.1 | 2 | 688 | 90 | ||
More Information
Set Membership: |
|

TRABD2A
Genomic Map