Assay Details
| Gene Symbol: MCMDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758967.3 | XP_002759013.1 | 11 - 12 | 2851 | 76 | |
| XM_008982738.2 | XP_008980986.1 | 10 - 11 | 2721 | 76 | ||
More Information
Set Membership: |
|

MCMDC2
Genomic Map