Assay Details
| Gene Symbol: OLFML1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754972.3 | XP_002755018.1 | 1 - 2 | 630 | 67 | |
| XM_017977874.1 | XP_017833363.1 | 1 - 2 | 633 | 67 | ||
More Information
Set Membership: |
|

OLFML1
Genomic Map