Assay Details
| Gene Symbol: ZNF652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748455.3 | XP_002748501.1 | 2 - 3 | 1399 | 82 | |
| XM_017972683.1 | XP_017828172.1 | 3 - 4 | 1338 | 82 | ||
| XM_017972684.1 | XP_017828173.1 | 3 - 4 | 1330 | 82 | ||
| XM_017972685.1 | XP_017828174.1 | 3 - 4 | 1333 | 82 | ||
More Information
Set Membership: |
|

ZNF652
Genomic Map