Assay Details
| Gene Symbol: KIAA0753 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003732980.3 | XP_003733028.1 | 7 - 8 | 2271 | 66 | |
| XM_008997284.2 | XP_008995532.1 | 7 - 8 | 1657 | 66 | ||
| XM_008997287.2 | XP_008995535.1 | 6 - 7 | 856 | 66 | ||
| XM_017972618.1 | XP_017828107.1 | 7 - 8 | 1657 | 66 | ||
More Information
Set Membership: |
|

KIAA0753
Genomic Map