Assay Details
| Gene Symbol: MB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743722.2 | XP_002743768.1 | 4 | 797 | 65 | |
| XM_009009915.1 | XP_009008163.1 | 4 | 670 | 65 | ||
| XM_009009918.2 | XP_009008166.1 | 3 | 575 | 65 | ||
More Information
Set Membership: |
|

MB
Genomic Map