Assay Details
| Gene Symbol: HEBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747085.3 | XP_002747131.1 | 2 - 3 | 523 | 80 | |
| XM_017971428.1 | XP_017826917.1 | 2 - 3 | 523 | 80 | ||
More Information
Set Membership: |
|

HEBP2
Genomic Map