Assay Details
| Gene Symbol: GABRA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744130.4 | XP_002744176.1 | 5 - 6 | 588 | 75 | |
| XM_008984539.2 | XP_008982787.1 | 6 - 7 | 821 | 75 | ||
More Information
Set Membership: |
|

GABRA1
Genomic Map