Assay Details
| Gene Symbol: TRIM17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760808.3 | XP_002760854.2 | 6 - 7 | 1000 | 93 | |
| XM_008985670.2 | XP_008983918.1 | 5 - 6 | 1099 | 93 | ||
| XM_008985671.2 | XP_008983919.1 | 5 - 6 | 2068 | 93 | ||
| XM_008985674.2 | XP_008983922.1 | 5 - 6 | 1345 | 93 | ||
More Information
Set Membership: |
|

TRIM17
Genomic Map