Assay Details
| Gene Symbol: LAYN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754375.4 | XP_002754421.1 | 6 - 7 | 1505 | 81 | |
| XM_009006760.2 | XP_009005008.1 | 6 - 7 | 1187 | 81 | ||
| XM_009006761.2 | XP_009005009.1 | 4 - 5 | 314 | 81 | ||
| XM_017977536.1 | XP_017833025.1 | 6 - 7 | 1693 | 81 | ||
More Information
Set Membership: |
|

LAYN
Genomic Map