Assay Details
| Gene Symbol: LAGE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002763427.4 | XP_002763473.1 | 2 - 3 | 686 | 57 | |
| XM_008990109.2 | XP_008988357.1 | 2 - 3 | 686 | 57 | ||
More Information
Set Membership: |
|

LAGE3
Genomic Map