Assay Details
Gene Symbol: ST6GALNAC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002743341.4 | XP_002743387.1 | 3 - 4 | 524 | 109 | |
XM_009003367.2 | XP_009001615.1 | 3 - 4 | 421 | 109 | ||
XM_009003376.2 | XP_009001624.1 | 3 - 4 | 529 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
